Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

755648

Sigma-Aldrich

β-Alanine-15N tert-butyl ester

98 atom % 15N, 98% (CP)

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C7H1515NO2
Molecular Weight:
146.19
MDL number:
UNSPSC Code:
12352108
PubChem Substance ID:
Pricing and availability is not currently available.

isotopic purity

98 atom % 15N

assay

98% (CP)

application(s)

peptide synthesis

mass shift

M+1

storage temp.

-10 to -25°C

SMILES string

CC(C)(C)OC(=O)CC[15NH2]

InChI

1S/C7H15NO2/c1-7(2,3)10-6(9)4-5-8/h4-5,8H2,1-3H3/i8+1

InChI key

ZJXHVYSDMUKUCA-VJJZLTLGSA-N

Packaging

This product may be available from bulk stock and can be packaged on demand. For information on pricing, availability and packaging, please contact Stable Isotopes Customer Service.

pictograms

Corrosion

signalword

Danger

hcodes

Hazard Classifications

Eye Dam. 1

Storage Class

11 - Combustible Solids

wgk_germany

WGK 3

flash_point_f

No data available

flash_point_c

No data available


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service