755400
7-Fluoro-5-iodoindole-3-carboxaldehyde
97%
Synonym(s):
7-Fluoro-5-iodo-1H-indole-3-carbaldehyde
About This Item
assay
97%
mp
244-249 °C
SMILES string
Fc1cc(I)cc2c(C=O)c[nH]c12
InChI
1S/C9H5FINO/c10-8-2-6(11)1-7-5(4-13)3-12-9(7)8/h1-4,12H
InChI key
KJHBVQBQLSUAQO-UHFFFAOYSA-N
signalword
Warning
hcodes
pcodes
Hazard Classifications
Eye Irrit. 2
Storage Class
13 - Non Combustible Solids
wgk_germany
WGK 3
flash_point_f
Not applicable
flash_point_c
Not applicable
Choose from one of the most recent versions:
Certificates of Analysis (COA)
It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.
If you need assistance, please contact Customer Support.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service