Skip to Content
MilliporeSigma
All Photos(2)

Key Documents

755346

Sigma-Aldrich

(4-Nitrobenzyl)mercaptan

95%

Synonym(s):

(4-Nitrophenyl)methanethiol, 4-Nitrobenzyl thiol

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C7H7NO2S
CAS Number:
Molecular Weight:
169.20
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

assay

95%

form

low-melting solid

mp

52-56 °C

SMILES string

[O-][N+](=O)c1ccc(CS)cc1

InChI

1S/C7H7NO2S/c9-8(10)7-3-1-6(5-11)2-4-7/h1-4,11H,5H2

InChI key

ACFHLWCEZHYYKX-UHFFFAOYSA-N

pictograms

Exclamation mark

signalword

Warning

hcodes

pcodes

Hazard Classifications

Skin Sens. 1

Storage Class

12 - Non Combustible Liquids

wgk_germany

WGK 3

flash_point_f

No data available

flash_point_c

No data available


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

T Watabe et al.
Biochemical and biophysical research communications, 131(2), 687-693 (1985-09-16)
4-Nitrobenzyl mercaptan (NBM) was enzymatically transformed at pH 6.0 into its S-sulfate in rat liver cytosol fortified with 3'-phosphoadenosine 5'-phosphosulfate. At pH 7.4, the S-sulfate was not detected from the incubation mixture. 4-Nitrobenzyl alcohol was also transformed under the same
Philip R Lindstedt et al.
Cell chemical biology, 28(1), 70-77 (2020-11-21)
Great advances have been made over the last four decades in therapeutic and diagnostic applications of antibodies. The activity maturation of antibody candidates, however, remains a significant challenge. To address this problem, we present a method that enables the systematic

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service