Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

754684

Sigma-Aldrich

5,7-Dibromoindole-3-carboxaldehyde

97%

Synonym(s):

5,7-Dibromo3-formylindole, 5,7-Dibromoindole-3-carbaldehyde

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C9H5Br2NO
CAS Number:
Molecular Weight:
302.95
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

assay

97%

form

solid

mp

200-205 °C

functional group

aldehyde
bromo

SMILES string

Brc1cc(Br)c2[nH]cc(C=O)c2c1

InChI

1S/C9H5Br2NO/c10-6-1-7-5(4-13)3-12-9(7)8(11)2-6/h1-4,12H

InChI key

JNRVKWFIKFDURF-UHFFFAOYSA-N

pictograms

Skull and crossbones

signalword

Danger

hcodes

Hazard Classifications

Acute Tox. 3 Oral

Storage Class

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

wgk_germany

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service