Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

754412

Sigma-Aldrich

N-Dodecyl-N-(2-pyridylmethylene)amine

97%

Synonym(s):

N-(n-Dodecyl)-N-(pyridin-2-ylmethylene)amine, N-Dodecyl-2-pyridylmethanimine, Dodecyl ATRP ligand

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C18H30N2
CAS Number:
Molecular Weight:
274.44
MDL number:
UNSPSC Code:
12162002
PubChem Substance ID:
NACRES:
NA.23

assay

97%

form

liquid

refractive index

n20/D 1.498

density

0.906 g/mL at 25 °C

SMILES string

CCCCCCCCCCCC\N=C\c1ccccn1

InChI

1S/C18H30N2/c1-2-3-4-5-6-7-8-9-10-12-15-19-17-18-14-11-13-16-20-18/h11,13-14,16-17H,2-10,12,15H2,1H3/b19-17+

InChI key

RJOCXVBEBNUEJB-HTXNQAPBSA-N

pictograms

Exclamation mark

signalword

Warning

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

target_organs

Respiratory system

Storage Class

10 - Combustible liquids

wgk_germany

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Perrier, S.; et al.
Macromolecules, 35 (8), 2941-2948 (2002)

Articles

Micro review of reversible addition/fragmentation chain transfer (RAFT) polymerization.

Protocols

We presents an article featuring procedures that describe polymerization of methyl methacrylate and vinyl acetate homopolymers and a block copolymer as performed by researchers at CSIRO.

We present an article about RAFT, or Reversible Addition/Fragmentation Chain Transfer, which is a form of living radical polymerization.

Polymerization via ATRP procedures demonstrated by Prof. Dave Haddleton's research group at the University of Warwick.

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service