Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

754269

Sigma-Aldrich

11-Mercaptoundecylphosphonic acid

95%

Synonym(s):

1-Thio-11-undecanephosphonic acid

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C11H25O3PS
CAS Number:
Molecular Weight:
268.35
MDL number:
UNSPSC Code:
12352103
PubChem Substance ID:
NACRES:
NA.23

assay

95%

form

solid

mp

80-90 °C

storage temp.

2-8°C

SMILES string

OP(O)(=O)CCCCCCCCCCCS

InChI

1S/C11H25O3PS/c12-15(13,14)10-8-6-4-2-1-3-5-7-9-11-16/h16H,1-11H2,(H2,12,13,14)

InChI key

VXLGFBKKMPMGST-UHFFFAOYSA-N

General description

11-Mercaptoundecylphosphonic acid (MUP) is an alkyl thiol that forms a self-assembled monolayer on a variety of surfaces. The thiol groups act as the anchor that facilitates the bonding of sulfur molecule to the surface atoms. The phosphoric and trifluoroacetate groups are useful molecules in the detection of monolayers.[1]

application

MUP can functionalize gold surface by forming a capped layer that can be used as a surface modified electrode material for the fabrication of sensors for biomedical and other industrial applications.[2][3][4]
Phosphonic acid that can be used to form self-assembled monolayers onto metal oxide surfaces. Also contains a thiol group that can be used to do click-type chemistry with alkenes or can bind metals.

pictograms

Exclamation mark

signalword

Warning

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

target_organs

Respiratory system

Storage Class

11 - Combustible Solids

wgk_germany

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Engineering Gram Selectivity of Mixed-Charge Gold Nanoparticles by Tuning the Balance of Surface Charges
Pillai PP, et al.
Angewandte Chemie (International ed. in English), 55(30), 8610-8614 (2016)
A omega-mercaptoundecylphosphonic acid chemically modified gold electrode for uranium determination in waters in presence of organic matter
Merli D, et al.
Talanta, 151(30), 119-125 (2016)
Karthik Rathinam et al.
Environmental science & technology, 53(9), 5202-5211 (2019-04-09)
Silica scaling of membranes used in reverse osmosis desalination processes is a severe problem, especially during the desalination of brackish groundwater due to high silica concentrations. This problem limits the water supply in inland arid and semiarid regions. Here, we
Phosphonic and sulfonic acid-functionalized gold nanoparticles: A solid-state NMR study
Fiurasek P and Reven L
Langmuir, 23(5), 2857-2866 (2007)
Quantification of silane molecules on oxidized silicon: are there options for a traceable and absolute determination?
Dietrich PM, et al.
Analytical Chemistry, 87(19), 10117-10124 (2015)

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service