Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

754250

Sigma-Aldrich

Cesium chromate

99.99% trace metals basis

Synonym(s):

Caesium chromate, Chromic acid cesium salt

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
Cs2CrO4
CAS Number:
Molecular Weight:
381.80
EC Number:
MDL number:
UNSPSC Code:
12352302
PubChem Substance ID:
NACRES:
NA.23
assay:
99.99% trace metals basis
form:
powder or crystals
Product 754250 is not currently sold in your country. Contact Technical Service

Quality Level

assay

99.99% trace metals basis

form

powder or crystals

impurities

≤150.0 ppm Trace Metal Analysis

SMILES string

[Cs+].[Cs+].[O-][Cr]([O-])(=O)=O

InChI

1S/Cr.2Cs.4O/q;2*+1;;;2*-1

InChI key

BROHICCPQMHYFY-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

signalword

Danger

Hazard Classifications

Aquatic Acute 1 - Aquatic Chronic 1 - Carc. 1B - Ox. Sol. 2 - Skin Sens. 1

Storage Class

5.1B - Oxidizing hazardous materials

wgk_germany

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Bender, J.; Wohlfarth, A.; Hoch, C.
Zeitschrift fur Naturforschung, B: Chemical Sciences, 65, 1416-1416 (2010)
Palekar; V.M.; Tierney; J.W.
Applied Catalysis A: General, 103, 105-105 (1993)

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service