Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

754021

Sigma-Aldrich

PSiF-DBT

Synonym(s):

Poly[2,7-(9,9-dioctyl-dibenzosilole)-alt-4,7-bis(thiophen-2-yl)benzo-2,1,3-thiadiazole], Poly[2,1,3-benzothiadiazole-4,7-diyl-2,5-thiophenediyl(9,9-dioctyl-9H-9-silafluorene-2,7-diyl)-2,5-thiophenediyl]

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
(C42H46N2S3Si)n
CAS Number:
UNSPSC Code:
12352103
NACRES:
NA.23

form

solid

mol wt

average Mn 10,000-80,000

loss

0.5 wt. %, 300 °C

mp

493-498 °C

orbital energy

HOMO 5.39 eV 

semiconductor properties

P-type (mobility=10−3 cm2/V·s)

Looking for similar products? Visit Product Comparison Guide

General description

PSiF-DBT is a conductive polymer that can be used as an active layer with thermal stability and high degree of π-electron delocalization. It has a high mobility of charge carrier around 1×10-3cm2/(V s) which can be useful in organic electronics.[1]

Application

PSiF-DBT can be used as a semiconducting polymer for the fabrication of polymeric solar cells (PSCs) and organic photovoltaic cells (OPVs).[2][3][4]

pictograms

Exclamation mark

signalword

Warning

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

target_organs

Respiratory system

Storage Class

11 - Combustible Solids

wgk_germany

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Toward high performance inverted polymer solar cells
Gong X, et al.
Polymer, 53(24), 5437-5448 (2012)
Annealing effect on donor-acceptor interface and its impact on the performance of organic photovoltaic devices based on PSiF-DBT copolymer and C60
Marchiori CFN, et al.
Applied Physics Letters, 106(13) (2015)
A direct route towards polymer/copper indium sulfide nanocomposite solar cells
Rath T, et al.
Advanced Energy Materials, 1(6), 1046-1050 (2011)
Electronic and structural properties in thermally annealed PSiF-DBT: PC71BM blends for organic photovoltaics
Borges BGAL, et al.
Thin Solid Films, 615(24), 165-170 (2016)
High-performance polymer heterojunction solar cells of a polysilafluorene derivative
Wang, E.; Wang, L.; Lan, L.; et al.
Applied Physics Letters, 92, 033307-033307 (2008)

Articles

The development of high-performance conjugated organic molecules and polymers has received widespread attention in industrial and academic research.

Organic materials in optoelectronic devices like LEDs and solar cells are of significant academic and commercial interest.

Thin, lightweight, and flexible electronic devices meet widespread demand for scalable, portable, and robust technology.

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service