Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

753904

Sigma-Aldrich

CdSeS/ZnS alloyed quantum dots kit

COOH functionalized, fluorescence λem 490-665 nm, 6 nm diameter, 5 X 1 mg/mL in H2O

Synonym(s):

CdSeS/ZnS alloyed quantum dots, QD carboxy functionalized, Fluorescent Nanocrystals, Trilite

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CdSxSe1-x/ZnS
UNSPSC Code:
26111700
NACRES:
NA.23

concentration

5 X 1 mg/mL in H2O

refractive index

n20/D 1.496 (lit.)

diameter

6 nm

bp

110-111 °C (lit.)

fluorescence

λem 490-665 nm

Storage temp.

2-8°C

Looking for similar products? Visit Product Comparison Guide

Legal Information

For use in optical electronics research only.
Trilite is a trademark of Cytodiagnostics Inc.

Pictograms

Environment

hcodes

Hazard Classifications

Aquatic Chronic 2

Storage Class

12 - Non Combustible Liquids

flash_point_f

Not applicable

flash_point_c

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Articles

Professor Sharma and colleagues review the synthesis and applications of this novel material. This includes a discussion of the unique properties of quantum dots and their suitability for solar cell applications, along with common synthesis techniques used to develop these materials.

Professor Xiaohu Gao (University of Washington, USA) provides a overview of recent quantum dot (QD) advancements and their potential for advancing bioassay and bioimaging technologies.

Perovskite quantum dots research progresses overcoming challenges, enabling rapid development of light-emitting devices.

Controlled synthesis of metal clusters regulates ligands and atoms, advancing metal nanomaterial synthesis.

See All

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service