753890
CdSeS/ZnS alloyed quantum dots
COOH functionalized, fluorescence λem 665 nm, 6 nm diameter, 1 mg/mL in H2O
Synonym(s):
QD carboxyl functionalized, Fluorescent Nanocrystals, Trilite™
About This Item
Recommended Products
Quality Level
form
liquid
concentration
1 mg/mL in H2O
refractive index
n20/D 1.496 (lit.)
diameter
6 nm
bp
110-111 °C (lit.)
fluorescence
λem 665 nm
storage temp.
2-8°C
Looking for similar products? Visit Product Comparison Guide
Application
Analysis Note
Legal Information
hcodes
pcodes
Hazard Classifications
Aquatic Chronic 2
Storage Class
12 - Non Combustible Liquids
wgk_germany
WGK 3
flash_point_f
Not applicable
flash_point_c
Not applicable
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Articles
Dye-sensitized solar cells as a promising, low-cost photovoltaic technology.
Professor Sharma and colleagues review the synthesis and applications of this novel material. This includes a discussion of the unique properties of quantum dots and their suitability for solar cell applications, along with common synthesis techniques used to develop these materials.
Professor Xiaohu Gao (University of Washington, USA) provides a overview of recent quantum dot (QD) advancements and their potential for advancing bioassay and bioimaging technologies.
Perovskite quantum dots research progresses overcoming challenges, enabling rapid development of light-emitting devices.
Active Filters
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service