Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

753866

Sigma-Aldrich

CdSeS/ZnS alloyed quantum dots

COOH functionalized, fluorescence λem 540 nm, 6 nm diameter, 1 mg/mL in H2O

Synonym(s):

QD carboxyl functionalized, Fluorescent Nanocrystals, Trilite

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CdSxSe1-x/ZnS
UNSPSC Code:
26111700
NACRES:
NA.23

Quality Level

form

liquid

composition

CdSeS/CdS/ZnS/ZnCdS

concentration

1 mg/mL in H2O

refractive index

n20/D 1.496 (lit.)

diameter

6 nm

bp

110-111 °C (lit.)

fluorescence

λem 540 nm

storage temp.

2-8°C

Looking for similar products? Visit Product Comparison Guide

General description

CdSeS/ZnS alloyed quantum dot (QD) is a quantum dot based semiconducting material that can be synthesized by mixing a CdSeS QDs solution with Zn(OAc)2 solution. Its luminescent properties make it useful in a variety of electronics and optoelectronics applications.[1][2]

Application

CdSeS/ZnS QDs can be used in the fabrication of organic light emitting diodes (OLEDs).[3] It may also be used in single dot imaging and other biological imaging.[4][5] It can potentially be used as a color converting light emitter for solid state lighting applications.[6]
These Photo and Thermostable Quantum Dot materials have comparable fluorescence and molecular kinetics across all colors. Narrow and sharp emission peaks result in less overlap of peaks when multiplexing.

Analysis Note

Full Width at Half Maximum: 30 - 40nm

Legal Information

For use in optical electronics research only.
Product of CytoDiagnostics
Trilite is a trademark of Cytodiagnostics Inc.

pictograms

Environment

hcodes

Hazard Classifications

Aquatic Chronic 2

Storage Class

12 - Non Combustible Liquids

wgk_germany

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

CdSeS/ZnS alloyed nanocrystal lifetime and blinking studies under electrochemical control
Qin W, et al.
ACS Nano, 6(1), 912-918 (2011)
Microwave-Assisted Aqueous Synthesis of Small-Sized, Highly Luminescent CdSeS/ZnS Core/Shell Quantum Dots for Live Cell Imaging
Zhan H, et al.
European Journal of Inorganic Chemistry, 2012(15), 2487-2493 (2012)
Fraser Stewart et al.
Scientific reports, 11(1), 2584-2584 (2021-01-30)
Biologic drugs, defined as therapeutic agents produced from or containing components of a living organism, are of growing importance to the pharmaceutical industry. Though oral delivery of medicine is convenient, biologics require invasive injections because of their poor bioavailability via
CdS x Se 1-x/ZnS semiconductor nanocrystal laser with sub 10kW/cm 2 threshold and 40nJ emission output at 600 nm
McLellan LJ, et al.
Optics Express, 24(2), A146-A153 (2016)
CdSe/CdS/ZnS and CdSe/ZnSe/ZnS Core- Shell- Shell Nanocrystals
Talapin DV, et al.
The Journal of Physical Chemistry B, 108(49), 18826-18831 (2004)

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service