Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

753823

Sigma-Aldrich

CdSeS/ZnS alloyed quantum dots kit

fluorescence λem 490-665 nm, 6 nm diameter, 5 X 1 mg/mL in toluene

Synonym(s):

CdSeS/ZnS alloyed quantum dots, Fluorescent Nanocrystals, Trilite

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CdSxSe1-x/ZnS
UNSPSC Code:
26111700
NACRES:
NA.23

description

Size: 1 ml x 5 vials

Quality Level

concentration

5 X 1 mg/mL in toluene

refractive index

n20/D 1.496 (lit.)

particle size

5.5-6.5 nm

diameter

6 nm

bp

110-111 °C (lit.)

fluorescence

λem 490+/-5 nm FWHM 30 nm in toluene (also available as 753750)
λem 490-665 nm
λem 525+/-5 nm FWHM 30 nm in toluene (also available as 753769)
λem 575+/-5 nm FWHM 30 nm in toluene (also available as 753785)
λem 630+/-5 nm FWHM 30 nm in toluene (also available as 753793)
λem 665+/-5 nm FWHM 30 nm in toluene (also available as 753807)

functional group

oleic acid

Storage temp.

2-8°C

Looking for similar products? Visit Product Comparison Guide

Legal Information

For use in optical electronics research only.
Trilite is a trademark of Cytodiagnostics Inc.

signalword

Danger

Hazard Classifications

Aquatic Chronic 2 - Asp. Tox. 1 - Flam. Liq. 2 - Repr. 2 - Skin Irrit. 2 - STOT RE 2 - STOT SE 3

target_organs

Central nervous system, Respiratory system

Storage Class

3 - Flammable liquids

wgk_germany

WGK 3

flash_point_f

39.2 °F - closed cup

flash_point_c

4.0 °C - closed cup


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service