Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

753750

Sigma-Aldrich

CdSeS/ZnS alloyed quantum dots

fluorescence λem 490 nm, 6 nm diameter, 1 mg/mL in toluene

Synonym(s):

QD alkyl functionalized, Fluorescent Nanocrystals, Trilite

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CdSxSe1-x/ZnS
UNSPSC Code:
26111700
NACRES:
NA.23

form

liquid

Quality Level

concentration

1 mg/mL in toluene

refractive index

n20/D 1.496 (lit.)

diameter

6 nm

bp

110-111 °C (lit.)

density

0.865 g/mL at 25 °C (lit.)

fluorescence

λem 490 nm

functional group

oleic acid

storage temp.

2-8°C

Looking for similar products? Visit Product Comparison Guide

Application

These Photo and Thermostable Quantum Dot materials have comparable fluorescence and molecular kinetics across all colors. Narrow and sharp emission peaks result in less overlap of peaks when multiplexing.

Analysis Note

Full Width at Half Maximum: 30 - 40nm

Legal Information

For use in optical electronics research only.
Product of CytoDiagnostics
Trilite is a trademark of Cytodiagnostics Inc.

signalword

Danger

Hazard Classifications

Aquatic Chronic 2 - Asp. Tox. 1 - Flam. Liq. 2 - Repr. 2 - Skin Irrit. 2 - STOT RE 2 - STOT SE 3

target_organs

Central nervous system

Storage Class

3 - Flammable liquids

wgk_germany

WGK 3

flash_point_f

39.2 °F

flash_point_c

4 °C


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service