Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

753513

Sigma-Aldrich

5-(2-Bromoethyl)-2,2-dimethyl-1,3-dioxane

95%

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C8H15BrO2
CAS Number:
Molecular Weight:
223.11
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:

assay

95%

form

liquid

refractive index

n20/D 1.482

density

1.336 g/mL at 25 °C

functional group

bromo
ether
ketal

storage temp.

−20°C

SMILES string

CC1(C)OCC(CCBr)CO1

InChI

1S/C8H15BrO2/c1-8(2)10-5-7(3-4-9)6-11-8/h7H,3-6H2,1-2H3

InChI key

QFLUXIUZRCNNHY-UHFFFAOYSA-N

pictograms

Exclamation mark

signalword

Warning

hcodes

Hazard Classifications

Acute Tox. 4 Oral - Eye Irrit. 2

Storage Class

10 - Combustible liquids

wgk_germany

WGK 3

flash_point_f

154.0 °F

flash_point_c

67.8 °C


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service