Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

753459

Sigma-Aldrich

Acetylene-1,2-diyl bis(boronic acid pinacol ester)

97%

Synonym(s):

1,2-Bis(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)ethyne, Ethyne-1,2-diyl bis(boronic acid pinacol ester), Ethyne-1,2-diyldiboronic acid dipinacol ester

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C14H24B2O4
CAS Number:
Molecular Weight:
277.96
MDL number:
UNSPSC Code:
12352103
PubChem Substance ID:

assay

97%

form

powder

mp

269 °C

storage temp.

2-8°C

SMILES string

CC1(C)OB(OC1(C)C)C#CB2OC(C)(C)C(C)(C)O2

InChI

1S/C14H24B2O4/c1-11(2)12(3,4)18-15(17-11)9-10-16-19-13(5,6)14(7,8)20-16/h1-8H3

InChI key

FABDBLONUBCKBB-UHFFFAOYSA-N

Application

Reactant for the preparation of:
  • Poly(p-phenyleneethynylene)s (PPEs), which are used in constructing organic light-emitting diodes (OLEDs), field-effect transistors (FETs) and several other materials used in electronic devices[1]

Storage Class

13 - Non Combustible Solids

wgk_germany

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Youn K Kang et al.
Organic letters, 10(7), 1341-1344 (2008-03-14)
We report the synthesis, structure, and characterization of a novel ethyne synthon, 1,2-bis(4',4',5',5'-tetramethyl[1',3',2']dioxaborolan-2'-yl)ethyne (B2C2). We demonstrate the utility of B2C2 in the Suzuki-Miyaura polycondensation reaction, synthesizing a water-soluble poly(p-phenyleneethynylene) from [2,5-diiodo-1,4-bis(3-propoxy-sulfonicacid)benzene] sodium salt in neat water under an aerobic atmosphere.

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service