Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

753335

Sigma-Aldrich

Chloro[2-(dicyclohexylphosphino)biphenyl]gold(I)

Synonym(s):

CyJohnPhos AuCl, Cyclohexyl JohnPhos AuCl

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C24H31AuClP
CAS Number:
Molecular Weight:
582.90
MDL number:
UNSPSC Code:
12352300
PubChem Substance ID:

form

solid

reaction suitability

core: gold
reagent type: catalyst

mp

241-244 °C

SMILES string

Cl[Au].C1CCC(CC1)P(C2CCCCC2)c3ccccc3-c4ccccc4

InChI

1S/C24H31P.Au.ClH/c1-4-12-20(13-5-1)23-18-10-11-19-24(23)25(21-14-6-2-7-15-21)22-16-8-3-9-17-22;;/h1,4-5,10-13,18-19,21-22H,2-3,6-9,14-17H2;;1H/q;+1;/p-1

InChI key

XDZKBEGPIOIGLI-UHFFFAOYSA-M

Storage Class

13 - Non Combustible Solids

wgk_germany

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service