Skip to Content
MilliporeSigma
All Photos(2)

Key Documents

752908

Sigma-Aldrich

2,2′-Bithieno[3,2-b]thiophene

96%

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C12H6S4
CAS Number:
Molecular Weight:
278.44
MDL number:
UNSPSC Code:
12352103
PubChem Substance ID:
NACRES:
NA.23

assay

96%

form

powder or crystals

mp

232-238 °C

SMILES string

C1(C=C(C2=CC3=C(C=CS3)S2)S4)=C4C=CS1

InChI

1S/C12H6S4/c1-3-13-9-5-11(15-7(1)9)12-6-10-8(16-12)2-4-14-10/h1-6H

InChI key

ZDFFDKMGCBNSKY-UHFFFAOYSA-N

application

This material is used as a binary pi-conjugated spacer in the synthesis of highly efficient; stable dyes for dye-sensitized solar cells.[1]

pictograms

Exclamation mark

signalword

Warning

hcodes

Hazard Classifications

Acute Tox. 4 Oral

Storage Class

11 - Combustible Solids

wgk_germany

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Guangliang Zhang et al.
Chemical communications (Cambridge, England), (16), 2198-2200 (2009-04-11)
We employed a binary spacer of orderly conjugated 3,4-ethyldioxythiophene and thienothiophene to construct a wide-spectral response organic chromophore for dye-sensitized solar cells, exhibiting a high power conversion efficiency of 9.8% measured under irradiation of 100 mW cm(-2) air mass 1.5

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service