Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

752665

Sigma-Aldrich

Aluminum zinc oxide

sputtering target, diam. × thickness 3.00 in. × 0.125 in., 99.99% trace metals basis

Synonym(s):

AZO

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
Al2O3/ZnO
CAS Number:
EC Number:
MDL number:
UNSPSC Code:
12352300
PubChem Substance ID:
NACRES:
NA.23

Quality Level

assay

99.99% trace metals basis

form

solid

composition

Aluminum oxide, 2 wt. %
Zinc oxide, 98 wt. %

reaction suitability

core: aluminum

diam. × thickness

3.00 in. × 0.125 in.

SMILES string

O=[Zn].O=[Al]O[Al]=O

InChI

1S/2Al.4O.Zn

InChI key

UQEOWMLGPTWJCL-UHFFFAOYSA-N

pictograms

Environment

signalword

Warning

hcodes

Hazard Classifications

Aquatic Acute 1 - Aquatic Chronic 1

Storage Class

11 - Combustible Solids

wgk_germany

WGK 2


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Serenelius, B. E.; Berggren, K. F.; Jin, Z. C.; Hamberg, I.; Grangvist, C. G.
Physical Review. B, Condensed Matter and Materials Physics, 37, 10244-10244 (1988)
Kim. K. H.; Park, K. C.; Ma, D. Y.
Journal of Applied Physics, 81, 7764-7764 (1997)

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service