Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

752657

Sigma-Aldrich

Indium tin oxide

sputtering target, diam. × thickness 3.00 in. × 0.125 in., 99.99% trace metals basis

Synonym(s):

ITO

Sign Into View Organizational & Contract Pricing


About This Item

CAS Number:
MDL number:
UNSPSC Code:
12352103
PubChem Substance ID:
NACRES:
NA.23

assay

99.99% trace metals basis

form

solid

diam. × thickness

3.00 in. × 0.125 in.

mp

1910 °C (lit.)

density

1.2 g/mL at 25 °C (lit.)

SMILES string

O=[SnH2].O=[In]O[In]=O.O=[In]O[In]=O.O=[In]O[In]=O.O=[In]O[In]=O.O=[In]O[In]=O.O=[In]O[In]=O.O=[In]O[In]=O.O=[In]O[In]=O.O=[In]O[In]=O

InChI

1S/2In.5O.Sn

InChI key

LNNWKAUHKIHCKO-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Storage Class

13 - Non Combustible Solids

wgk_germany

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Shigesato, Y.; Takaki, S.; Haranoh, T.
Journal of Applied Physics, 71, 3356-3356 (1992)
Kim, H.; Pique, A.; Horwitz, J. S.; Mattoussi, H.; Murata, H.; Karari, Z. H.; Chrisey, D. B.
Applied Physics Letters, 74, 3444-3444 (1999)

Articles

Nanocomposite Coatings with Tunable Properties Prepared by Atomic Layer Deposition

Continuous efficiency improvements in photovoltaic devices result from material advancements and manufacturing innovation.

Combinatorial Materials Science identifies breakthrough materials through systematic exploration, aiding material discovery.

The properties of many devices are limited by the intrinsic properties of the materials that compose them.

See All

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service