Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

752606

Sigma-Aldrich

N-Butyl-2-pyridylmethanimine

97%

Synonym(s):

N-(2-Pyridinylmethylene)-1-butanamine, Butyl ATRP ligand

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C10H14N2
CAS Number:
Molecular Weight:
162.23
MDL number:
UNSPSC Code:
12162002
PubChem Substance ID:
NACRES:
NA.23

assay

97%

form

liquid

refractive index

n20/D 1.519

density

0.949 g/mL at 25 °C

SMILES string

CCCC\N=C\c1ccccn1

InChI

1S/C10H14N2/c1-2-3-7-11-9-10-6-4-5-8-12-10/h4-6,8-9H,2-3,7H2,1H3/b11-9+

InChI key

YLHWKRVTRIJWAB-PKNBQFBNSA-N

Storage Class

10 - Combustible liquids

wgk_germany

WGK 3

flash_point_f

228.9 °F

flash_point_c

109.4 °C


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

ATRP for Everyone: Ligands and Initiators for the Clean Synthesis of Functional Polymers
Jakubowski, W.; Tsarevsky, N.V.; McCarthy, P., Matyjaszewski, K.
Material Matters, 5.1, 16-16 (2010)
Larisa V Sigolaeva et al.
Polymers, 12(7) (2020-07-12)
We demonstrate the application of amphiphilic ionic poly(n-butylmethacrylate)-block- poly(2-(dimethylamino)ethyl methacrylate) diblock copolymers (PnBMA40-b-PDMAEMA40, PnBMA40-b-PDMAEMA120, PnBMA70-b-PDMAEMA120) for dispersing multiwalled carbon nanotubes (MWCNTs) in aqueous media, a subsequent efficient surface modification of screen-printed electrodes (SPEs), and the application of the modified SPEs
Perrier, S.; et al.
Macromolecules, 35 (8), 2941-2948 (2002)

Articles

Micro review of reversible addition/fragmentation chain transfer (RAFT) polymerization.

Protocols

We presents an article featuring procedures that describe polymerization of methyl methacrylate and vinyl acetate homopolymers and a block copolymer as performed by researchers at CSIRO.

We present an article about RAFT, or Reversible Addition/Fragmentation Chain Transfer, which is a form of living radical polymerization.

Polymerization via ATRP procedures demonstrated by Prof. Dave Haddleton's research group at the University of Warwick.

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service