Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

752495

Sigma-Aldrich

Poly(ethylene glycol) methyl ether (2-methyl-2-propionic acid dodecyl trithiocarbonate)

average Mn 10,000

Synonym(s):

Poly(ethylene glycol) methyl ether [2-(dodecylthiocarbonothioylthio)-2-methylpropionate]

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12162002
NACRES:
NA.23

form

solid

mol wt

average Mn 10,000

transition temp

Tm 57-62 °C

PDI

≤1.1

storage temp.

2-8°C

General description

Need help choosing the correct RAFT Agent? Please consult the RAFT Agent to Monomer compatibility table.

Molecular weight: PEG average Mn = 10 kDa; n ~227

Application

RAFT agent for controlled radical polymerization; especially suited for the polymerization of styrene; acrylate; and acrylamide monomers to make lithographically and biologically important PEG-block copolymers. Chain Transfer Agent (CTA)

Storage Class

13 - Non Combustible Solids

wgk_germany

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

RAFT Agent Design and Synthesis
Keddie, D. J.; et al.
Macromolecules, 45, 5321-5342 (2012)
Massimo Benaglia et al.
Journal of the American Chemical Society, 131(20), 6914-6915 (2009-05-01)
The polymerization of most monomers that are polymerizable by radical polymerization can be controlled by the reversible addition-fragmentation chain transfer (RAFT) process. However, it is usually required that the RAFT agent be selected according to the types of monomer being

Articles

Reversible addition–fragmentation chain transfer (RAFT) polymerization is rapidly moving to the forefront in construction of drug and gene delivery vehicles.

Over the past two decades, the rapid advance of controlled living polymerization (CLP) techniques.

The modification of biomacromolecules, such as peptides and proteins, through the attachment of synthetic polymers has led to a new family of highly advanced biomaterials with enhanced properties.

Micro review of reversible addition/fragmentation chain transfer (RAFT) polymerization.

See All

Protocols

We present an article about RAFT, or Reversible Addition/Fragmentation Chain Transfer, which is a form of living radical polymerization.

We presents an article featuring procedures that describe polymerization of methyl methacrylate and vinyl acetate homopolymers and a block copolymer as performed by researchers at CSIRO.

Polymerization via ATRP procedures demonstrated by Prof. Dave Haddleton's research group at the University of Warwick.

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service