Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

752487

Sigma-Aldrich

Poly(ethylene glycol) methyl ether (4-cyano-4-pentanoate dodecyl trithiocarbonate)

average Mn 1,400

Synonym(s):

Dodecyl-trithiocarbonate-cyano PEG, PEG-RAFT, Poly(ethylene glycol)methyl ether 4-cyano-4-[(dodecylsulfanylthiocarbonyl)sulfanyl]pentanoate

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352100
NACRES:
NA.23

form

solid

Quality Level

mol wt

average Mn 1,400

reaction suitability

reagent type: chemical modification reagent
reaction type: Polymerization Reactions

transition temp

Tm 27-33 °C

PDI

≤1.1

Ω-end

4-cyano-4-pentanoate dodecyl trithiocarbonate

α-end

methoxy

polymer architecture

shape: linear
functionality: monofunctional

storage temp.

2-8°C

Looking for similar products? Visit Product Comparison Guide

General description

Need help choosing the correct RAFT Agent? Please consult the RAFT Agent to Monomer compatibility table.

Molecular weight: PEG average Mn 1,000 Da; n ~22

Application

RAFT agent for controlled radical polymerization; especially suited for the polymerization of styrene, acrylate, and acrylamide monomers to make lithographically and biologically important PEG-block copolymers. Chain Transfer Agent (CTA)

Storage Class

13 - Non Combustible Solids

wgk_germany

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

RAFT Agent Design and Synthesis
Keddie, D. J.; et al.
Macromolecules, 45, 5321-5342 (2012)

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service