Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

752363

Sigma-Aldrich

Poly(ethylene glycol), 16 hydroxyl dendron, generation 3

average Mn 21,600

Synonym(s):

PFD-G3-PEG20k-OH, Dendron functionalized PEG, generation 3

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12162002
NACRES:
NA.23

description

hydroxyl surface groups

form

solid

mol wt

PEG average Mn 20,000
average Mn 21,600
generation 3

no. Surface Groups

16

mp

56-60 °C

functional group

hydroxyl

Looking for similar products? Visit Product Comparison Guide

Other Notes

average PEG Mn = 21,600; esterified with 16 OH groups

Storage Class

11 - Combustible Solids

wgk_germany

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Qigang Wang et al.
Nature, 463(7279), 339-343 (2010-01-22)
With the world's focus on reducing our dependency on fossil-fuel energy, the scientific community can investigate new plastic materials that are much less dependent on petroleum than are conventional plastics. Given increasing environmental issues, the idea of replacing plastics with

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service