Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

752215

Sigma-Aldrich

Vinyl valerate

contains ≤1000 ppm hydroquinone as stabilizer, 97%

Synonym(s):

Pentanoic acid, ethenyl ester, Valeric acid, vinyl ester, Vinyl pentanoate

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C7H12O2
CAS Number:
Molecular Weight:
128.17
MDL number:
UNSPSC Code:
12162002
PubChem Substance ID:
NACRES:
NA.23

assay

97%

form

liquid

contains

≤1000 ppm hydroquinone as stabilizer

refractive index

n20/D 1.417

density

0.899 g/mL at 25 °C

storage temp.

−20°C

SMILES string

CCCCC(=O)OC=C

InChI

1S/C7H12O2/c1-3-5-6-7(8)9-4-2/h4H,2-3,5-6H2,1H3

InChI key

BLZSRIYYOIZLJL-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

pictograms

FlameEnvironment

signalword

Warning

hcodes

Hazard Classifications

Aquatic Acute 1 - Aquatic Chronic 3 - Flam. Liq. 3

Storage Class

3 - Flammable liquids

wgk_germany

WGK 3

flash_point_f

93.9 °F - closed cup

flash_point_c

34.4 °C - closed cup


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Slide 1 of 1

1 of 1

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service