Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

752177

Sigma-Aldrich

Benzyl diazoacetate

contains ~10% dichloromethane as stabilizer

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C9H8N2O2
CAS Number:
Molecular Weight:
176.17
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

form

liquid

contains

~10% dichloromethane as stabilizer

availability

available only in USA

refractive index

n20/D 1.551

density

1.171 g/mL at 25 °C

storage temp.

2-8°C

SMILES string

[N-]=[N+]=CC(=O)OCc1ccccc1

InChI

1S/C9H8N2O2/c10-11-6-9(12)13-7-8-4-2-1-3-5-8/h1-6H,7H2

InChI key

JBBKWIFIXPBQGZ-UHFFFAOYSA-N

signalword

Danger

Hazard Classifications

Acute Tox. 4 Oral - Carc. 2 - Eye Irrit. 2 - Self-react. C - Skin Irrit. 2 - STOT SE 3

target_organs

Respiratory system

Storage Class

5.2 - Organic peroxides and self-reacting hazardous materials

wgk_germany

WGK 3


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service