Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

751952

Sigma-Aldrich

Sodium bromide hydrate

99.999% trace metals basis

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
NaBr·xH2O
Molecular Weight:
102.89 (anhydrous basis)
MDL number:
UNSPSC Code:
12352302
PubChem Substance ID:

assay

99.999% trace metals basis

form

solid

composition

Br, 88.5-91.2% silver nitrate titration

impurities

≤15.0 ppm Trace Metal Analysis

SMILES string

[Na+].[Br-].O

InChI

1S/BrH.Na.H2O/h1H;;1H2/q;+1;/p-1

InChI key

NEPUNMOZAQLFSU-UHFFFAOYSA-M

Application

Sodium Bromide has been used as a reactant in the green synthesis of aryl bromides(1), as well as serving as a molten flux for the growth of inorganic crystals(2).

Storage Class

11 - Combustible Solids

wgk_germany

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Schmidt, R.; et al.
Green Chemistry, 14, 1673-1679 (2012)
Shivakumara, C.; et al.
Mat. Res. Bul., 39, 71-81 (2004)

Articles

The diversity of applications and nanostructured materials accessible using ultrasonic spray methods are highlighted in this article.

Ultrasonic spray pyrolysis produces scalable nanomaterials like metal oxides and quantum dots for diverse applications.

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service