Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

751928

Sigma-Aldrich

Zirconium(IV) bromide

anhydrous, 99.99% trace metals basis

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
ZrBr4
CAS Number:
Molecular Weight:
410.84
EC Number:
MDL number:
UNSPSC Code:
12352302
PubChem Substance ID:

grade

anhydrous

assay

99.99% trace metals basis

form

solid

mp

≥450 °C (lit.)

application(s)

battery manufacturing

SMILES string

Br[Zr](Br)(Br)Br

InChI

1S/4BrH.Zr/h4*1H;/q;;;;+4/p-4

InChI key

LSWWNKUULMMMIL-UHFFFAOYSA-J

Looking for similar products? Visit Product Comparison Guide

Application

Zirconium bromide has been used as a starting material in the synthesis of zirconium hydride complexes[1] as well as unique cluster compounds[2].

pictograms

Corrosion

signalword

Danger

hcodes

Hazard Classifications

Skin Corr. 1B

supp_hazards

Storage Class

8B - Non-combustible corrosive hazardous materials

wgk_germany

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Qi, R.Y.; Corbett, J.D.
Inorganic Chemistry, 37, 1646-1651 (1995)
Lee, H.; et al.
Journal of the American Chemical Society, 120, 5830-5831 (1998)

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service