Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

751847

Sigma-Aldrich

1,2-Bis(2,2-diethoxyethyl) disulfide

technical grade, 85%

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C12H26O4S2
CAS Number:
Molecular Weight:
298.46
MDL number:
UNSPSC Code:
12352001
PubChem Substance ID:
NACRES:
NA.22

grade

technical grade

assay

85%

form

liquid

refractive index

n20/D 1.478

density

1.036 g/mL at 25 °C

SMILES string

CCOC(CSSCC(OCC)OCC)OCC

InChI

1S/C12H26O4S2/c1-5-13-11(14-6-2)9-17-18-10-12(15-7-3)16-8-4/h11-12H,5-10H2,1-4H3

InChI key

XJIKEAQNTPCOLU-UHFFFAOYSA-N

Storage Class

12 - Non Combustible Liquids

wgk_germany

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service