Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

751480

Sigma-Aldrich

Malonic acid

sublimed grade, ≥99.95% trace metals basis

Synonym(s):

Propanedioic acid

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH2(COOH)2
CAS Number:
Molecular Weight:
104.06
Beilstein/REAXYS Number:
1751370
EC Number:
MDL number:
UNSPSC Code:
12352300
PubChem Substance ID:

grade

sublimed grade

assay

≥99.95% trace metals basis

form

powder

mp

132-135 °C (dec.) (lit.)

SMILES string

OC(=O)CC(O)=O

InChI

1S/C3H4O4/c4-2(5)1-3(6)7/h1H2,(H,4,5)(H,6,7)

InChI key

OFOBLEOULBTSOW-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

The common technique for producing malonic acid is from chloroacetic acid.[1]Malonic acid undergoes thermal decarboxylation at temperatures of 135-137oC.2

pictograms

Corrosion

signalword

Danger

hcodes

Hazard Classifications

Eye Dam. 1

Storage Class

13 - Non Combustible Solids

wgk_germany

WGK 1

flash_point_f

314.6 °F - closed cup

flash_point_c

157 °C - closed cup


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Organic Syntheses, 18, 50-50 (1938)
E A Haan et al.
European journal of pediatrics, 144(6), 567-570 (1986-04-01)
A second child with a more severe deficiency of malonyl CoA decarboxylase is described. He is mildly mentally retarded and presented with vomiting, a seizure, hypoglycaemia and mild metabolic acidosis during a urinary tract infection. The urine contained increased amounts
Bogdan Belgorodsky et al.
Bioconjugate chemistry, 16(5), 1058-1062 (2005-09-22)
The preparation and characterization of the stable human serum albumin (HSA)-C3 isomer of tris-malonic acid [C60]fullerene complex is reported. Other than the anti-fullerene antibody, a stable protein-fullerene complex with a native protein has never been observed. This study may provide
Arturo Orellana et al.
The Journal of organic chemistry, 77(12), 5286-5296 (2012-05-04)
An efficient synthesis of (-)-kainic acid, through a high-pressure-promoted Diels-Alder cycloaddition of a vinylogous malonate derived from 4-hydroxyproline, is described. The bicyclic adduct could be converted into the natural product with complete stereocontrol.
Adriana Wilhelm et al.
Journal of medicinal chemistry, 55(22), 9817-9830 (2012-10-31)
The protein kinase C-related kinase 2 (PRK2)-interacting fragment (PIF) pocket of phosphoinositide-dependent kinase-1 (PDK1) was proposed as a novel target site for allosteric modulators. In the present work, we describe the design, synthesis, and structure-activity relationship of a series of

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service