Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

751308

Sigma-Aldrich

4-Aminopiperidine-3,3,4,5,5-d5

98 atom % D, 98% (CP)

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C5D5H7N2
Molecular Weight:
105.19
MDL number:
UNSPSC Code:
12352005
PubChem Substance ID:

isotopic purity

98 atom % D

assay

98% (CP)

density

0.991 g/mL at 25 °C

mass shift

M+5

SMILES string

[2H]C1([2H])CNCC([2H])([2H])C1([2H])N

InChI

1S/C5H12N2/c6-5-1-3-7-4-2-5/h5,7H,1-4,6H2/i1D2,2D2,5D

InChI key

BCIIMDOZSUCSEN-QJWYSIDNSA-N

Packaging

This product may be available from bulk stock and can be packaged on demand. For information on pricing, availability and packaging, please contact Stable Isotopes Customer Service.

pictograms

FlameCorrosion

signalword

Danger

hcodes

Hazard Classifications

Eye Dam. 1 - Flam. Liq. 3 - Skin Corr. 1B

Storage Class

3 - Flammable liquids

wgk_germany

WGK 3

flash_point_f

114.1 °F - closed cup

flash_point_c

45.6 °C - closed cup


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service