Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

751197

Sigma-Aldrich

3,5-Difluoroethylbenzene

97%

Synonym(s):

1-Ethyl-3,5-difluorobenzene, 5-Ethyl-1,3-difluorobenzene

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C8H8F2
CAS Number:
Molecular Weight:
142.15
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

assay

97%

form

liquid

refractive index

n20/D 1.452

density

1.065 g/mL at 25 °C

functional group

fluoro

SMILES string

CCc1cc(F)cc(F)c1

InChI

1S/C8H8F2/c1-2-6-3-7(9)5-8(10)4-6/h3-5H,2H2,1H3

Inchi Key

PFINYKRPUJCMCT-UHFFFAOYSA-N

Pictograms

FlameExclamation mark

signalword

Warning

hcodes

Hazard Classifications

Acute Tox. 4 Oral - Flam. Liq. 3

Storage Class

3 - Flammable liquids

wgk_germany

WGK 3

flash_point_f

88.0 °F

flash_point_c

31.1 °C


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service