Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

751103

Sigma-Aldrich

Potassium indole-5-trifluoroborate

96%

Synonym(s):

Potassium 5-indolyltrifluoroborate

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C8H6BF3KN
CAS Number:
Molecular Weight:
223.04
MDL number:
UNSPSC Code:
12352103
PubChem Substance ID:

assay

96%

form

solid

mp

173-181 °C

SMILES string

[K+].F[B-](F)(F)c1ccc2[nH]ccc2c1

InChI

1S/C8H6BF3N.K/c10-9(11,12)7-1-2-8-6(5-7)3-4-13-8;/h1-5,13H;/q-1;+1

InChI key

BLUZPWXLGGDSRM-UHFFFAOYSA-N

pictograms

Exclamation mark

signalword

Warning

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

target_organs

Respiratory system

Storage Class

11 - Combustible Solids

wgk_germany

WGK 2

flash_point_f

Not applicable

flash_point_c

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service