Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

750875

Sigma-Aldrich

6-Hydroxyhexanoic acid-13C6

99 atom % 13C, 95% (CP)

Synonym(s):

6-Hydroxycaproic acid-13C6

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
HO(13CH2)513COOH
Molecular Weight:
138.11
MDL number:
UNSPSC Code:
51113400
PubChem Substance ID:
Pricing and availability is not currently available.

isotopic purity

99 atom % 13C

assay

95% (CP)

mp

50-52 °C

mass shift

M+6

storage temp.

−20°C

SMILES string

O[13CH2][13CH2][13CH2][13CH2][13CH2][13C](O)=O

InChI

1S/C6H12O3/c7-5-3-1-2-4-6(8)9/h7H,1-5H2,(H,8,9)/i1+1,2+1,3+1,4+1,5+1,6+1

InChI key

IWHLYPDWHHPVAA-IDEBNGHGSA-N

Packaging

This product may be available from bulk stock and can be packaged on demand. For information on pricing, availability and packaging, please contact Stable Isotopes Customer Service.

Storage Class

11 - Combustible Solids

wgk_germany

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service