Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

750409

Sigma-Aldrich

(R)-(+)-2-(Methoxymethoxy)-1-propanol

Synonym(s):

MOM protected (R)-1,2-Propanediol

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C5H12O3
CAS Number:
Molecular Weight:
120.15
MDL number:
UNSPSC Code:
12352001
PubChem Substance ID:
NACRES:
NA.22

form

liquid

optical activity

[α]/D +77.8°, c = 0.5% in chloroform

refractive index

n20/D 1.415

density

1.002 g/mL at 25 °C

functional group

ether
hydroxyl

SMILES string

COCO[C@H](C)CO

InChI

1S/C5H12O3/c1-5(3-6)8-4-7-2/h5-6H,3-4H2,1-2H3/t5-/m1/s1

InChI key

YNDXFEKPXUXDQS-RXMQYKEDSA-N

Looking for similar products? Visit Product Comparison Guide

pictograms

Exclamation mark

signalword

Warning

Hazard Classifications

Acute Tox. 4 Oral - Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

target_organs

Respiratory system

Storage Class

10 - Combustible liquids

wgk_germany

WGK 3

flash_point_f

167.9 °F

flash_point_c

75.5 °C


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service