750204
(3-Ammoniopropyl)di-tert-butylphosphonium bis(tetrafluoroborate)
96%
About This Item
Recommended Products
assay
96%
form
solid
reaction suitability
reaction type: Buchwald-Hartwig Cross Coupling Reaction
reaction type: Heck Reaction
reaction type: Hiyama Coupling
reaction type: Negishi Coupling
reaction type: Sonogashira Coupling
reaction type: Stille Coupling
reaction type: Suzuki-Miyaura Coupling
reagent type: ligand
mp
304-310 °C
functional group
phosphine
SMILES string
F[B-](F)(F)F.F[B-](F)(F)F.CC(C)(C)[PH+](CCC[NH3+])C(C)(C)C
InChI
1S/C11H26NP.2BF4/c1-10(2,3)13(9-7-8-12)11(4,5)6;2*2-1(3,4)5/h7-9,12H2,1-6H3;;/q;2*-1/p+2
InChI key
YFPVMYMPAJSJHB-UHFFFAOYSA-P
Storage Class
11 - Combustible Solids
wgk_germany
WGK 3
flash_point_f
Not applicable
flash_point_c
Not applicable
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Don't see the Right Version?
If you require a particular version, you can look up a specific certificate by the Lot or Batch number.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service