Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

750077

Sigma-Aldrich

Thieno[2,3-b]thiophene-2,5-dicarboxaldehyde

97% (GC)

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C8H4O2S2
CAS Number:
Molecular Weight:
196.25
MDL number:
UNSPSC Code:
12352103
PubChem Substance ID:

assay

97% (GC)

form

powder or crystals

mp

185-190 °C

SMILES string

O=Cc1cc2cc(C=O)sc2s1

InChI

1S/C8H4O2S2/c9-3-6-1-5-2-7(4-10)12-8(5)11-6/h1-4H

InChI key

BYRWPDKQHOEMTA-UHFFFAOYSA-N

Application

This material can be used to create materials that generate stable diradicals[1] useful for electronics applications and potentially some Organic Photovoltaic Applications.

pictograms

Exclamation mark

signalword

Warning

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - Skin Sens. 1 - STOT SE 3

target_organs

Respiratory system

Storage Class

11 - Combustible Solids

wgk_germany

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Akita, T.; et al.
Tetrahedron Letters, 36, 5543-5546 (1995)

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service