Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

749788

Sigma-Aldrich

Indium tin oxide coated PET

surface resistivity 250 Ω/sq, L × W × thickness 1 ft × 1 ft × 7 mil, sheet

Synonym(s):

ITO-PET, Polyethylene terephthalate film, ITO coated

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
In2O3 · (SnO2)x
MDL number:
UNSPSC Code:
12352103
PubChem Substance ID:
NACRES:
NA.23

form

sheet

surface resistivity

250 Ω/sq

L × W × thickness

1 ft × 1 ft × 7 mil

SMILES string

O=[In]O[In]=O.O=[Sn]=O

InChI

1S/2In.5O.Sn

InChI key

LNNWKAUHKIHCKO-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Indium tin oxide coated polyethylene terephthalate (PET) film is optically transparent and electrically conductive.The ITO coating is protected by a thin, transparent film, that can be easily peeled off.

Application

ITO-PET may be used in fabricating flexible OLEDs. ITO/PET may be used as a reference anode in conventional flexible polymer based organic solar cells (OSCs).

Features and Benefits

The polyester film products comprise sputtered ITO, and exhibit transmittance and resistance characteristics similar to those of the glass substrates.
These polyester film products comprise of sputtered ITO, and exhibit transmittance and resistance characteristics similar to those of glass substrates.

Physical properties

Thickness of ITO coating is 1000 Å

Storage Class

13 - Non Combustible Solids

wgk_germany

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

High-Performance Organic Light-Emitting Diodes Using ITO Anodes Grown on Plastic by Room-Temperature Ion-Assisted Deposition
Yang Y, et al.
Advanced Materials, 16(4), 321-324 (2004)
Modular mass spectrometric tool for analysis of composition and phosphorylation of protein complexes
Blethrow JD, et al.
PLoS ONE, 2(4), e358-e358 (2007)
Efficient and Flexible ITO-Free Organic Solar Cells Using Highly Conductive Polymer Anodes.
Na SI, et al.
Advanced Materials, 20(21), 4061-4067 (2008)

Articles

A transparent conductive electrode (TCE) is an essential component of various optoelectronic devices such as solar cells, liquid-crystal displays (LCD), light-emitting diodes (LED), and touch screens.

Progress in solution-processed functional materials leads to thin-film optoelectronic devices for industrial and consumer electronics.

Recent research highlights tunable properties of inorganic nanoparticles, driving interest in optoelectronics.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service