749680
Propylthiol functionalized silica
mesoporous, nanoparticles, TCP-porphine labeled, 200 nm particle size, pore size 4 nm
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
UNSPSC Code:
12352302
form
mesoporous
nanoparticles
contains
meso-Tetra(4-carboxyphenyl)porphine as fluorescent label
particle size
200 nm
pore size
4 nm pore size
bp
2230 °C (lit.)
mp
1600-1725 °C (lit.)
Looking for similar products? Visit Product Comparison Guide
General description
These well defined mesoporous materials are used in applications that include separations, drug delivery, optical imaging, and composites.
Storage Class
13 - Non Combustible Solids
wgk_germany
WGK 3
flash_point_f
Not applicable
flash_point_c
Not applicable
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Lot/Batch Number
Don't see the Right Version?
If you require a particular version, you can look up a specific certificate by the Lot or Batch number.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service