Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

749664

Sigma-Aldrich

Propylcarboxylic acid functionalized silica

mesoporous, nanoparticles, 200 nm particle size, pore size 4 nm

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352302
NACRES:
NA.23

form

mesoporous
nanoparticles

particle size

200 nm

pore size

4 nm pore size

bp

2230 °C (lit.)

mp

1600-1725 °C (lit.)

Looking for similar products? Visit Product Comparison Guide

pictograms

Exclamation mark

signalword

Warning

hcodes

Hazard Classifications

Eye Irrit. 2 - STOT SE 3

target_organs

Respiratory system

Storage Class

10 - Combustible liquids

wgk_germany

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Articles

Mesoporous Materials include a range of high surface area porous silicates with applications in gas adsorption, drug delivery, diagnostics and catalysis.

Mesoporous silica used in wastewater remediation, air cleaning, catalysis, bio-catalysis, and drug delivery.

Silica's versatility spans various industries, including biomedical applications.

The recent emergence of a number of highly functional nanomaterials has enabled new approaches to the understanding, diagnosis, and treatment of cancer.

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service