Skip to Content
MilliporeSigma
All Photos(2)

Key Documents

749583

Sigma-Aldrich

Piperidine-4-carbonitrile

97%

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C6H10N2
CAS Number:
Molecular Weight:
110.16
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:

assay

97%

form

liquid

refractive index

n20/D 1.476

density

0.987 g/mL at 25 °C

functional group

nitrile

storage temp.

2-8°C

SMILES string

N#CC1CCNCC1

InChI

1S/C6H10N2/c7-5-6-1-3-8-4-2-6/h6,8H,1-4H2

InChI key

FSDNTQSJGHSJBG-UHFFFAOYSA-N

pictograms

CorrosionExclamation mark

signalword

Danger

hcodes

Hazard Classifications

Acute Tox. 4 Oral - Eye Dam. 1

Storage Class

10 - Combustible liquids

wgk_germany

WGK 3

flash_point_f

221.0 °F

flash_point_c

105 °C


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Man Zhang et al.
Heterocycles, 101(1), 145-164 (2020-08-11)
Neuropathic pain, epilepsy, insomnia, and tremor disorder may arrive from an increase of intracellular Ca2+ concentration through a dysfunction of T-type Ca2+ channels. Thus, T-type calcium channels could be a target in drug discovery for the treatments of neuropathic pain

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service