Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

749575

Sigma-Aldrich

4-aminomethyl quinoline hydrochloride

Synonym(s):

4-Quinolinemethanamine hydrochloride

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C10H10N2 · HCl
CAS Number:
Molecular Weight:
194.66
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

form

solid

mp

205-208 °C (decomposition)

functional group

amine

SMILES string

Cl.NCc1ccnc2ccccc12

InChI

1S/C10H10N2.ClH/c11-7-8-5-6-12-10-4-2-1-3-9(8)10;/h1-6H,7,11H2;1H

InChI key

WTPUWOBAIWBPGP-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

pictograms

Exclamation mark

signalword

Warning

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

target_organs

Respiratory system

Storage Class

11 - Combustible Solids

wgk_germany

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service