Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

749451

Sigma-Aldrich

1-Bromo-2-(1,1-difluoroethyl)benzene

97%

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C8H7BrF2
CAS Number:
Molecular Weight:
221.04
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

assay

97%

form

liquid

refractive index

n20/D 1.510

density

1.518 g/mL at 25 °C

functional group

bromo
fluoro

SMILES string

CC(F)(F)c1ccccc1Br

InChI

1S/C8H7BrF2/c1-8(10,11)6-4-2-3-5-7(6)9/h2-5H,1H3

InChI key

XZJWDIHNMMLGLA-UHFFFAOYSA-N

pictograms

Exclamation mark

signalword

Warning

hcodes

Hazard Classifications

Eye Irrit. 2

Storage Class

11 - Combustible Solids

wgk_germany

WGK 3

flash_point_f

197.1 °F

flash_point_c

91.7 °C


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service