Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

749184

Sigma-Aldrich

Poly(styrene-block-methyl methacrylate)

average Mn 30,000, block ratio of 1:1

Synonym(s):

PS-block-PMMA, PS-PMMA

Sign Into View Organizational & Contract Pricing


About This Item

CAS Number:
UNSPSC Code:
12162002
NACRES:
NA.23

form

pellets

mol wt

average Mn 15,000 (PMMA)
average Mn 15,000 (polystyrene)
average Mn 30,000

transition temp

Tm 342-347 °C

Mw/Mn

<1.2

InChI

1S/C8H8.C5H8O2/c1-2-8-6-4-3-5-7-8;1-4(2)5(6)7-3/h2-7H,1H2;1H2,2-3H3

InChI key

ADFPJHOAARPYLP-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

15 kDa PS; 15 kDa PMMA; m ~143; n ~150
PS Mn = 15 ± 2 kg/mol
PMMA Mn = 15 ± 2 kg/mol
PDI of the overall block copolymer <1.2
PS molecular weight verified by SEC (taking an aliquot from the reaction mixture) and the PMMA molecular weight by NMR spectroscopy. The overall block copolymer molecular weight distribution would be determined by SEC.

pictograms

Exclamation mark

signalword

Warning

hcodes

Hazard Classifications

Eye Irrit. 2

Storage Class

13 - Non Combustible Solids

wgk_germany

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

MN: Surgical LPN files product liability suit: cement Mfgr.'s failure to warn of health hazard.
Nursing law's Regan report, 42(8), 3-3 (2004-08-11)
M E Forsythe et al.
Journal of orthopaedic surgery (Hong Kong), 14(1), 38-42 (2006-04-07)
To compare the pharmacokinetic profile of tobramycin in blood, urine, and at the operative site following the use of Simplex-tobramycin bone cement in primary total hip replacement between patients with and without renal dysfunction. Six patients with renal dysfunction underwent
N S Dar-Odeh et al.
Journal of oral rehabilitation, 24(10), 755-760 (1997-12-31)
The aim of this study was to compare some of the physical properties of a heat-cured, a self-cured and a visible light-cured acrylic resin, and to evaluate the suitability of visible light-cured resin as a repair material for dentures made
G N Thomopoulos et al.
The journal of histochemistry and cytochemistry : official journal of the Histochemistry Society, 31(7), 871-878 (1983-07-01)
The high iron diamine (HID) method for detection of sulfated complex carbohydrate has been applied directly on thin sections of variably fixed tissues embedded in epoxy and nonepoxy resins. Results with postembedment HID staining in mouse intestinal epithelium are compared
Novel fabrication of 2D and 3D inverted opals and their application.
Jae Ah Lee et al.
Small (Weinheim an der Bergstrasse, Germany), 7(18), 2581-2586 (2011-08-03)

Articles

The development of drugs that target specific locations within the human body remains one of the greatest challenges in biomedicine today.

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service