Skip to Content
MilliporeSigma
All Photos(2)

Key Documents

749168

Sigma-Aldrich

Zirconium(IV) oxo hydroxy methacrylate

Synonym(s):

Hexameric Zr cluster, Tetra-μ3-hydroxyundecakis [μ-(2-methyl-2-propenoato-κO:κ;O′)](2-methyl-2-propenoato-κO)tetra-μ3-oxohexazirconium

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C48H64O32Zr6
CAS Number:
Molecular Weight:
1700.35
MDL number:
UNSPSC Code:
26111700
PubChem Substance ID:
NACRES:
NA.23

form

powder

Quality Level

mp

>300 °C (lit.)

SMILES string

O.O.CC(=C)C(=O)O[Zr-]1(=O)O\C(=[O+]\[Zr-](=O)(OC(=O)C(C)=C)O\C(=[O+]\[Zr-](=O)(OC(=O)C(C)=C)O\C(=[O+]\[Zr-](=O)(OC(=O)C(C)=C)O\C(=[O+]\[Zr-](=O)(OC(=O)C(C)=C)O\C(=[O+]\[Zr-](=O)(OC(=O)C(C)=C)O\C(=[O+]\1)C(C)=C)C(C)=C)C(C)=C)C(C)=C)C(C)=C)C(C)=C

InChI

1S/12C4H6O2.2H2O.6O.6Zr/c12*1-3(2)4(5)6;;;;;;;;;;;;;;/h12*1H2,2H3,(H,5,6);2*1H2;;;;;;;;;;;;/q;;;;;;;;;;;;;;;;;;;;6*+2/p-12

InChI key

DOFDBWIWCJHDLU-UHFFFAOYSA-B

pictograms

Exclamation mark

signalword

Warning

hcodes

Hazard Classifications

Skin Irrit. 2 - Skin Sens. 1

Storage Class

11 - Combustible Solids

wgk_germany

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Steffen Hausdorf et al.
Dalton transactions (Cambridge, England : 2003), 7(7), 1107-1113 (2009-03-27)
An ethynylene diisophthalic acid linker molecule was synthesized and used to form a zinc carboxylate-based metal organic framework (MOF) with very large pores and unit cell volume resulting from the unusual combination of structurally different inorganic units forming the secondary
Steffen Hausdorf et al.
Journal of the American Chemical Society, 132(32), 10978-10981 (2010-08-12)
A simple two-component procedure was developed to synthesize not only classical zinc-based IRMOFs represented by MOF-5 but also the cobalt and beryllium homologues of this most prominent MOF. The procedure is the first manifestation of mirroring the IRMOF series with
Steffen Hausdorf et al.
The journal of physical chemistry. A, 112(33), 7567-7576 (2008-07-26)
The contributions of terephthalic acid and Zn(2+)-coordinated water in N,N-diethylformamide (DEF) to the overall proton activity in the synthesis of MOF-5 (Zn4O(BDC)3, BDC = 1,4-benzenedicarboxylate) were quantitatively determined by combined electrochemical and UV-vis spectroscopic measurements. Structural transformations of zinc carboxylate-based

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service