Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

749109

Sigma-Aldrich

2-Fluorobenzaldehyde dimethyl acetal

97%

Synonym(s):

1-(Dimethoxymethyl)-2-fluorobenzene

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C9H11FO2
CAS Number:
Molecular Weight:
170.18
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:

assay

97%

form

liquid

density

1.105 g/mL at 25 °C

SMILES string

COC(OC)c1ccccc1F

InChI

1S/C9H11FO2/c1-11-9(12-2)7-5-3-4-6-8(7)10/h3-6,9H,1-2H3

InChI key

GHRDHBIBFZLUJM-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

pictograms

Exclamation mark

signalword

Warning

hcodes

Hazard Classifications

Acute Tox. 4 Oral

Storage Class

10 - Combustible liquids

wgk_germany

WGK 3

flash_point_f

163.0 °F

flash_point_c

72.8 °C


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service