Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

749028

Sigma-Aldrich

3-Ethyl-2-13C-3-pentanol-1,3,5-13C3

99 atom % 13C, 97% (CP)

Synonym(s):

Triethylcarbinol-(ethyl-2-13C-pentanol-1,3,5-13C3)

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
13C4C3H16O
Molecular Weight:
120.17
MDL number:
UNSPSC Code:
12352001
PubChem Substance ID:
Pricing and availability is not currently available.

isotopic purity

99 atom % 13C

assay

97% (CP)

bp

141 °C/743 mmHg (lit.)

density

0.852 g/mL at 25 °C

mass shift

M+4

SMILES string

[13CH3]C[13C](O)(C[13CH3])C[13CH3]

InChI

1S/C7H16O/c1-4-7(8,5-2)6-3/h8H,4-6H2,1-3H3/i1+1,2+1,3+1,7+1

InChI key

XKIRHOWVQWCYBT-LGKVPWJVSA-N

Packaging

This product may be available from bulk stock and can be packaged on demand. For information on pricing, availability and packaging, please contact Stable Isotopes Customer Service.

signalword

Danger

Hazard Classifications

Acute Tox. 4 Oral - Eye Dam. 1 - Flam. Liq. 3 - Skin Irrit. 2 - STOT SE 3

Storage Class

3 - Flammable Liquids

wgk_germany

WGK 3

flash_point_f

100.4 °F - closed cup

flash_point_c

38 °C - closed cup


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service