Skip to Content
MilliporeSigma
All Photos(2)

Key Documents

748889

Sigma-Aldrich

9-Fluorenylmethylthiol

97%

Synonym(s):

9H-Fluorene-9-methanethiol, FmSH

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C14H12S
CAS Number:
Molecular Weight:
212.31
MDL number:
UNSPSC Code:
12352103
PubChem Substance ID:
NACRES:
NA.23

assay

97%

form

solid

mp

31-36 °C

SMILES string

SCC1c2ccccc2-c3ccccc13

InChI

1S/C14H12S/c15-9-14-12-7-3-1-5-10(12)11-6-2-4-8-13(11)14/h1-8,14-15H,9H2

InChI key

MXHVORUHWLKKMZ-UHFFFAOYSA-N

Application

This novel material is capable of self-assembly onto gold surfaces. The fluorene group is a precursor for the synthesis of luminophore materials that have longer lifespans and more efficient luminescence.[1] The methylene group provides flexibility and a potential point of chirality.

pictograms

Environment

signalword

Warning

hcodes

pcodes

Hazard Classifications

Aquatic Acute 1

Storage Class

13 - Non Combustible Solids

wgk_germany

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Cheol, H. C.; Joo, S. H.; Kim, K.; Jin, J. I.; Shin, H. W.; Kim, Y. R.
Macromolecules, 38, 6336-6336 (2005)

Articles

Recent research highlights tunable properties of inorganic nanoparticles, driving interest in optoelectronics.

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service