Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

748595

Sigma-Aldrich

(1S,6S)-2,8-Diazabicyclo[4.3.0]nonane

97%

Synonym(s):

(4aS,7aS)-Octahydro-1H-pyrrolo[3,4-b]pyridine

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C7H14N2
CAS Number:
Molecular Weight:
126.20
MDL number:
UNSPSC Code:
12352005
PubChem Substance ID:

assay

97%

form

liquid

optical activity

[α]22/D −4.0°, c = 0.5 in chloroform

refractive index

n20/D 1.515

density

1.024 g/mL at 25 °C

storage temp.

2-8°C

SMILES string

C1CN[C@@H]2CNC[C@@H]2C1

InChI

1S/C7H14N2/c1-2-6-4-8-5-7(6)9-3-1/h6-9H,1-5H2/t6-,7+/m0/s1

InChI key

KSCPLKVBWDOSAI-NKWVEPMBSA-N

pictograms

Corrosion

signalword

Danger

hcodes

Hazard Classifications

Eye Dam. 1

Storage Class

10 - Combustible liquids

wgk_germany

WGK 2

flash_point_f

196.0 °F

flash_point_c

91.1 °C


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service