Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

748552

Sigma-Aldrich

N-Boc-α-(phenylsulfonyl)-4-methoxybenzylamine

95%

Synonym(s):

N-[(4-Methoxyphenyl)(phenylsulfonyl)methyl]carbamic acid 1,1-dimethylethyl ester, tert-Butyl[(phenylsulfonyl)(4-methoxyphenyl)methyl]carbamate, [(4-Methoxyphenyl)(phenylsulfonyl) methyl]carbamic acid tert-butyl ester

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C19H23NO5S
CAS Number:
Molecular Weight:
377.45
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

assay

95%

form

solid

mp

159-164 °C

functional group

amine
sulfone

storage temp.

2-8°C

SMILES string

COc1ccc(cc1)C(NC(=O)OC(C)(C)C)S(=O)(=O)c2ccccc2

InChI

1S/C19H23NO5S/c1-19(2,3)25-18(21)20-17(14-10-12-15(24-4)13-11-14)26(22,23)16-8-6-5-7-9-16/h5-13,17H,1-4H3,(H,20,21)

InChI key

JBOXLVHIHXWCPQ-UHFFFAOYSA-N

pictograms

Exclamation mark

signalword

Warning

hcodes

Hazard Classifications

Eye Irrit. 2

Storage Class

11 - Combustible Solids

wgk_germany

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service