Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

748390

Sigma-Aldrich

3,3-Dichloro-1,1,1,5,5,5-hexamethyltrisiloxane

Synonym(s):

Bis(trimethylsiloxy)dichlorosilane

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C6H18Cl2OSi3
CAS Number:
Molecular Weight:
261.37
MDL number:
UNSPSC Code:
12352103
PubChem Substance ID:

form

liquid

refractive index

n20/D 1.398

density

0.995 g/mL at 25 °C

Storage temp.

2-8°C

SMILES string

C[Si](C)(C)O[Si](Cl)(Cl)O[Si](C)(C)C

InChI

1S/C6H18Cl2O2Si3/c1-11(2,3)9-13(7,8)10-12(4,5)6/h1-6H3

Inchi Key

YQPFMTIZDAWFAC-UHFFFAOYSA-N

Pictograms

FlameCorrosion

signalword

Danger

hcodes

Hazard Classifications

Flam. Liq. 3 - Skin Corr. 1B

supp_hazards

Storage Class

3 - Flammable liquids

wgk_germany

WGK 3

flash_point_f

129.9 °F

flash_point_c

54.4 °C


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service